Learn More
ZEOprep™ NH2P Functionalized
Tienda Zeochem Productos
Click to view available options
:
1 kg
100 g
5 kg
500 g
Descripción
- NH2P perfect for ion exchange applications
- Irregular Amorphous Silica Gel
- High Purity
- European Manufacture
- Swiss accuracy and consistency
- Excellent batch to batch reproducibility
- ISO Certification
- High Chemical and Mechanical Stability
- Narrow Particles Size Distribution
- 20 to 500 μm particles size range
- Available in 60 A or 90 A pore size
- High loadability
- Selection of pack sizes available
- Suitable for gravity, flash, prep and process scale
Especificaciones
Especificaciones
| Tamaño de partículas | 40-63 μm |
| Tamaño de poro | 60 Å |
| Técnica | Chromatography |
| Cantidad | 5 kg |
| Tipo | Gel de sílice |
| Material | Gel de sílice |
| Fase | NH2P funcionalizado |
| Forma de partícula | Irregular |