Learn More
ZC3HAV1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. p-7208208 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16816574 20 μL 20 microlitros
16806574 150 μL 150 microlitros
2 options
Código de producto. 16816574
Marca: Proteintech 168201AP20UL
Rabbit Polyclonal Antibody
The zinc finger antiviral protein (ZC3HAV1) is a CCCH type zinc finger protein that induces an innate immunity to infections by retrovirus by preventing the accumulation of viral RNAs in the cytoplasm and recruits the RNA processing exosome to degrade target RNAs, thereby inhibiting virus replication. ZC3HAV1 is localized in the cytoplasm at steady state, but shuttles between nucleus and cytoplasm in a XPO1-dependent manner. ZAP is a direct target gene of IRF3 action in cellular antiviral responses.
Especificaciones
Antígeno ZC3HAV1
Aplicaciones Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.37 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica ZC3HAV1
N.º de referencia del gen Q3UPF5, Q7Z2W4, Q8K3Y6
Alias de gen FLB6421, ZAP, ZC3H2, ZC3HAV1, ZC3HDC2, Zinc finger antiviral protein
Símbolos de los genes ZC3HAV1
Especie del huésped Rabbit
Inmunógeno ZC3HAV1 Fusion Protein Ag10364
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 252832, 56829, 78781
Especies diana Mouse, Human, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG