Learn More
ZBTB2 Rabbit, anti-Human, Polyclonal, Proteintech
Código de producto. 18064505 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18064505 150 μL 150 microlitros
18019675 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18064505 Proveedor Proteintech N.º de proveedor 264361AP150UL
Rabbit Polyclonal Antibody
The ZBTB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZBTB2. This antibody recognizes human antigen. The ZBTB2 antibody has been validated for the following applications: IP, ELISA analysis.
Especificaciones
Antígeno ZBTB2
Aplicaciones Immunoprecipitation, ELISA
Concentración 350 μg/mL
Símbolos de los genes KIAA1483, ZBTB2, ZNF437
Especie del huésped Rabbit
Cantidad 150 μL
Primario o secundario Primary
ID de gen (Entrez) 57621
Especies diana Human
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG