Learn More
YWHAB Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Código de producto. 18040696 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
18040696 100 μL 100 microlitros
18016865 20 μL 20 microlitros
2 options
Código de producto. 18040696
Marca: Proteintech CL48810936100UL
Rabbit Polyclonal Antibody
The YWHAB antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human YWHAB. This antibody recognizes human, mouse, rat antigen. The YWHAB antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Especificaciones
Antígeno YWHAB
Aplicaciones Immunofluorescence
Concentración 1000 μg/mL
Conjugado CoraLite Plus 488
Símbolos de los genes 14 3 3 protein beta/alpha, 14 3 3,, GW128, HS1, KCIP 1, Protein 1054, YWHAB
Especie del huésped Rabbit
Cantidad 100 μL
Primario o secundario Primary
ID de gen (Entrez) 7529
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG