missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YOD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA535042
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with chicken and zebrafish based on sequence homology.
Deubiquitinating enzymes (DUBs) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA8 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
Especificaciones
| YOD1 | |
| Polyclonal | |
| Unconjugated | |
| YOD1 | |
| 9930028C20Rik; DUBA8; DUBA-8; fe01a10; HIN7; HIN-7; HIV-1-induced protease 7; HIV-induced protein-7-like protease; hsHIN7; hypothetical protein LOC550411; OTU domain containing 2; OTU domain-containing protein 2; OTUD2; PRO0907; Ubiquitin thioesterase OTU1; wu:fe01a10; Yod1; YOD1 deubiquitinase; YOD1 OTU deubiquinating enzyme 1 homolog; YOD1 OTU deubiquinating enzyme 1 homolog ( yeast); YOD1 OTU deubiquitinating enzyme 1 homologue; YOD1 OTU deubiquitinating enzyme 1 homologue (S. cerevisiae); zgc:112182 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 55432 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5VVQ6 | |
| YOD1 | |
| KLH conjugated synthetic peptide between 319-347 amino acids from the C-terminal region of human YOD1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido