missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YLPM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA561641
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: RDRGELRIRE YPERGDTWRE KRDYVPDRMD WERERLSDRW YPSDVDRHSP MAEHMPSSHH SSEMMGSDAS LDSDQGLGGV MVLSQRQHEI ILKA Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.
Plays a role in the reduction of telomerase activity during differentiation of embryonic stem cells by binding to the core promoter of TERT and controlling its down-regulation.
Especificaciones
| YLPM1 | |
| Polyclonal | |
| Unconjugated | |
| YLPM1 | |
| C14orf170; Nuclear protein ZAP3; PPP1R169; protein phosphatase 1, regulatory subunit 169; YLP motif containing 1; YLP motif-containing protein 1; YLPM1; ZAP; ZAP113; ZAP3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56252 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49750 | |
| YLPM1 | |
| Recombinant protein corresponding to Human YLPM1. Recombinant protein control fragment (Product #RP-98404). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido