Learn More
Invitrogen™ YIPF3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5113335
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
YIPF3 Polyclonal Antibody for Western Blot, ELISA
Involved in the maintenance of the Golgi structure. May play a role in hematopoiesis.
Especificaciones
| YIPF3 | |
| Polyclonal | |
| Unconjugated | |
| YIPF3 | |
| C6orf109; D17Wsu94e; dJ337H4.3; FinGER3; Killer lineage protein 1; KLIP1; Liver regeneration-related protein LRRGT00110; LRRGT00110; natural killer cell-specific antigen KLIP1; Protein YIPF3; Protein YIPF3, 36 kDa form III; Protein YIPF3, N-terminally processed; RGD1308185; Yip1 domain family member 3; Yip1 domain family, member 3; YIP1 family member 3; YIPF3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 25844, 28064 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q3UDR8, Q9GZM5 | |
| YIPF3 | |
| Recombinant Human Protein YIPF3 protein (2-148AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |