Learn More
Melford Yeast without Amino Acids
Código de producto. 30018947
Change view
Click to view available options
Cantidad:
250 g
500 g
1 kg
Tamaño de la unidad:
1 Kg
250 g
500 g
30018947 500 g 500 g
30018946 250 g 250 g
30018945 1 kg 1 Kg
3 options
Código de producto. 30018947
Marca: Melford Y20040500.0
For classifying yeasts based on amino acid and carbohydrates requirements.
For classifying yeasts based on amino acid and carbohydrates requirements. Contains all essential vitamins and inorganic salts necessary for the cultivaiton of yeasts. Excludes histidine methionine, tryptophan and carbon source. Also contains <1 mg/L each of vitamins and trace inorganic salts.
Especificaciones
Cantidad 500 g