Learn More
Melford Yeast Extract, Granulated
Código de producto. 30018938
Click to view available options
Cantidad:
500 g
1 kg
2 kg
5 kg
Tamaño de la unidad:
1 Kg
2 Kg
5 Kg
500 g
Código de producto. 30018938
Marca: Melford Y200252000.0
Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins.
Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins. Material is specially granulated to reduce dust and increase solubility rate. Suitable for Microbiology Applications.
Especificaciones
Cantidad 2 kg