Learn More
Melford Yeast with Amino Acids
Código de producto. 30018944
Change view
Click to view available options
Cantidad:
250 g
500 g
Tamaño de la unidad:
250 g
500 g
30018944 500 g 500 g
30018943 250 g 250 g
2 options
Código de producto. 30018944
Proveedor: Melford Y20030500.0
For classifying yeasts based on amino acid and carbohydrates requirements.
For classifying yeasts based on amino acid and carbohydrates requirements. Contains all essential vitamins and inorganic salts necessary for the cultivation of yeasts. Excludes carbohydrate source.
Especificaciones
Cantidad 500 g