Learn More
YAP1 Mouse anti-Human, Unconjugated, Clone: 3A7A9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66900-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The YAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human YAP1. This antibody recognizes human, mouse, rat antigen. The YAP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
| YAP1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
| 3A7A9 | |
| Unconjugated | |
| 20 μL | |
| 10413 | |
| IgG1 |