missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YAF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA530359
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: 293T, Raji. Predicted reactivity: Chicken (86%), Rhesus Monkey (86%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene interacts with YY1, a zinc finger protein involved in negative regulation of muscle-restricted genes. This gene product itself contains a single N-terminal C2-X10-C2 zinc finger, and in contrast to YY1, is up-regulated during myogenic differentiation. It also facilitates proteolytic cleavage of YY1 by the calcium- activated protease, m-calpain, suggesting a mechanism by which this protein antagonizes the negative effect of YY1.
Especificaciones
| YAF2 | |
| Polyclonal | |
| Unconjugated | |
| YAF2 | |
| 2810021M11Rik; MGC41856; YAF2; YY1 associated factor 2; YY1-associated factor 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10138 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8IY57 | |
| YAF2 | |
| Recombinant protein encompassing a sequence within the center region of human YAF2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido