missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XYLB Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA558144
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: DSQHYMALLC FKNGSLMREK IRNESVSRSW SDFSKALQST EMGNGGNLGF YFDVMEITPE IIGRHRFNTE NHKVAAFPGD V Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.
Human xylulokinase-like protein (XYLB), is a 58 kDa protein belongs to a family of enzymes that include fucokinase, gluconokinase, glycerokinase and xylulokinase. XYLB is a key regulator for controlling metabolic disease. XYLB also plays important roles in energy metabolism. XYLB has 2isoformsproduced byalternative splicing with the MW of 58 and 43 kDa.
Especificaciones
| XYLB | |
| Polyclonal | |
| Unconjugated | |
| XYLB | |
| AI132223; AI327421; D-xylulokinase; Xylb; Xylulokinase; xylulokinase homolog (H. influenzae); xylulose kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 102448, 9942 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75191, Q3TNA1 | |
| XYLB | |
| Recombinant protein corresponding to Human XYLB. Recombinant protein control fragment (Product #RP-96321). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido