Learn More
Chemsplash Xtreme 50 SMS T5/6 Coverall with BlueTaped Seams
Descripción
Chemsplash Xtreme SMS 50 Coverall
- 50GSM Breathable SMS Fabric
- Fully Taped Seams
- Elasticated 3 Pce Hood
- Elasticated Cuffs & Ankles
- Two Way Zip With Adhesive Zip Flap
- Silicone and Latex Free
- PFAS Free
- Comes without the chest label
Description: The Chemsplash Xtreme SMS 50 Coverall with Taped Seams is made from breathable 50gsm SMS fabric, compliant to Cat III Type 5 and 6. It offers a robust and strong barrier to dry hazards and is especially suitable for prolonged periods of use due to its breathable properties. All seams are bound.
Especificaciones
Especificaciones
| Tipo de producto | Hooded Coveralls |
| Tamaño | Small |
| Sexo | Unisex |
| Material | Tela de SMS |
| Color | Azul |
| Desechables | Single-use |
| Estilo de cintura | Elástico |
| Estilo de tobillo | Elastic |
| Estilo de manguito | Elastic |
| Estilo de cuello | Hooded |
| Mostrar más |