missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPR1 Polyclonal Antibody

Descripción
Recommended positive controls: A431, H1299, HepG2. Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Xenotropic and polytropic retrovirus receptor (XPR1) is an Orphan-U GPCR with an unknown ligand. ESTs have been isolated from brain, breast, kidney, pancreas, placenta, prostate, skeletal muscle, testis, and tonsil libraries.
Especificaciones
Especificaciones
| Antígeno | XPR1 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| génica | Xpr1 |
| N.º de referencia del gen | Q9UBH6 |
| Alias de gen | IBGC6; Protein SYG1 homolog; receptor for MCF virus 1; Rmc1; Rmc-1; RP11-46A10.1; suppressor of yeast G>a> deletion; susceptibility to xenotropic virus; Sxv; Syg1; X3; xenotropic and polytropic murine leukemia virus receptor; xenotropic and polytropic murine leukemia virus receptor X3; Xenotropic and polytropic retrovirus receptor 1; xenotropic and polytropic retrovirus receptor 1 homolog; XPR1; XR; X-receptor |
| Símbolos de los genes | Xpr1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?