missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA534338
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
XPR1 Polyclonal Antibody for ICC/IF, IHC (P)
Xenotropic and polytropic retrovirus receptor (XPR1) is an Orphan-U GPCR with an unknown ligand. ESTs have been isolated from brain, breast, kidney, pancreas, placenta, prostate, skeletal muscle, testis, and tonsil libraries.
Especificaciones
| XPR1 | |
| Polyclonal | |
| Unconjugated | |
| Xpr1 | |
| IBGC6; Protein SYG1 homolog; receptor for MCF virus 1; Rmc1; Rmc-1; RP11-46A10.1; suppressor of yeast G>a> deletion; susceptibility to xenotropic virus; Sxv; Syg1; X3; xenotropic and polytropic murine leukemia virus receptor; xenotropic and polytropic murine leukemia virus receptor X3; Xenotropic and polytropic retrovirus receptor 1; xenotropic and polytropic retrovirus receptor 1 homolog; XPR1; XR; X-receptor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100051159, 100339244, 100526125, 100689082, 19775, 289424, 424415, 490301, 536550, 9213 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9QZ70, Q9UBH6, Q9Z0U0 | |
| Xpr1 | |
| Synthetic 16 amino acid peptide from the N-terminus cytoplasmic domain of human XPR1. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Chicken, Bat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido