missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: DLSRDRPLVL LLHLLQLGPL FRCFEVFCIY FQSGNNEEPY VSITKKRQMP KNGLSEEIEK EVGQAEGKLI THRSAFSRA Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
The XK protein is a membrane-bound protein associated with the endoplasmic reticulum (ER) and forms part of the ER membrane complex (EMC), which is critical for various cellular processes. This protein is involved in the insertion, folding, and assembly of membrane proteins in eukaryotic cells. The XK protein's role in the ER highlights its importance in maintaining proper cell function, as the ER is integral in protein synthesis, lipid metabolism, and calcium storage. Furthermore, the XK protein's involvement in these processes underscores its significance in cellular homeostasis and its potential contribution to pathologies when mutated or dysfunctional.
Especificaciones
Especificaciones
| Antígeno | XK |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | XK |
| N.º de referencia del gen | P51811 |
| Alias de gen | 1810038K19Rik; Kell blood group precursor (McLeod phenotype); Kell blood group precursor (McLeod phenotype) homolog; kell complex 37 kDa component; KX; Kx antigen; MCLDS; McLeod syndrome gene homolog; membrane transport protein XK; NA; NAC; neuroacanthocytosis; neurocanthocytosis; X1k; Xk; XK homolog; XK, Kell blood group complex subunit (McLeod syndrome); Xkh; XKR1; XK-related protein 1; X-linked Kx blood group; X-linked Kx blood group (McLeod syndrome); X-linked Kx blood group (McLeod syndrome) homolog; Xrg1 |
| Símbolos de los genes | XK |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
