Learn More
XDH Mouse anti-Human, Clone: OTI1C10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF812777
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
XDH belongs to the group of molybdenum-containing hydroxylases involved in the oxidative metabolism of purines. The enzyme is a homodimer. This protein can be converted to xanthine oxidase by reversible sulfhydryl oxidation or by irreversible proteolytic modification. Defects in xanthine dehydrogenase cause xanthinuria, may contribute to adult respiratory stress syndrome, and may potentiate influenza infection through an oxygen metabolite-dependent mechanism.Especificaciones
| XDH | |
| Monoclonal | |
| Unconjugated | |
| XDH | |
| XAN1, XO, XOR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7498 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1C10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P47989 | |
| XDH | |
| Human recombit protein fragment corresponding to amino acids 1-280 of human XDH produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.