missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: TYRPRPRRSL QPPELIGAML EPTDEEPK Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 47%.
This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in normal testis, and in Ewing's sarcoma, rhabdomyosarcoma, a breast cancer and a germ cell tumor. The protein encoded by this gene shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens.
Especificaciones
Especificaciones
| Antígeno | XAGE2 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | XAGE2 |
| N.º de referencia del gen | Q96GT9 |
| Alias de gen | Cancer/testis antigen 12.2; cancer/testis antigen family 12, member 2; CT12.2; G antigen family D member 3; GAGED3; X antigen family member 2; X antigen family, member 2; X antigen family, member 2B; XAGE2; XAGE-2; XAGE2B |
| Símbolos de los genes | XAGE2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
