missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XAB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA14266
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PA1-4266 detects the XAB2 (XPA-binding protein 2) protein in human samples. PA1-4266 has been successfully used in Western blot procedures. PA1-4266 immunizing peptide corresponds to amino acid residues 840-855 of the human XAB2 protein.
A novel protein designated XAB2 (XPA-binding protein 2) interacts with XPA, a factor central to both nucleotide excision repair subpathways. Data suggests that XAB2 is a novel component involved in transcription-coupled repair and transcription.
Especificaciones
| XAB2 | |
| Polyclonal | |
| Unconjugated | |
| XAB2 | |
| 0610041O14Rik; Adapter protein ATH-55; Ath55; AV025587; crn-related protein kim1; DKFZp762C1015; HCNP; HCNP protein; HCRN; KIAA1177; NTC90; PP3898; Pre-mRNA-splicing factor SYF1; Protein HCNP; SYF1; SYF1 homolog, RNA splicing factor; SYF1 pre-mRNA-splicing factor; Xab2; XPA binding protein 2; XPA-binding protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56949 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9HCS7 | |
| XAB2 | |
| Synthetic peptide: E(840) Q Q S V P A A V F G S L K E D(855). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido