Learn More
TERUMO SR STRAIGHT PUR VERSATUS™ Intravenous Catheters
Código de producto. 17192734
Change view
Click to view available options
Calibre de aguja:
14 G
16 G
18 G
20 G
22 G
24 G
Longitud (métrico):
19 mm
25 mm
32 mm
51 mm
64 mm
Tamaño de la unidad:
paquete de 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Calibre de aguja Longitud (métrico) unitSize
17192734 20 G 32 mm paquete de 50
17152734 14 G 64 mm paquete de 50
17162734 16 G 51 mm paquete de 50
17172734 18 G 32 mm paquete de 50
17182734 18 G 51 mm paquete de 50
17102744 20 G 51 mm paquete de 50
17112744 22 G 25 mm paquete de 50
17122744 24 G 19 mm paquete de 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Código de producto. 17192734 Proveedor TERUMO N.º de proveedor 1SROF2032
Short PIVC with an internal needle.
  • The Versatus™ I.V. Catheters are made out of medical grade polyurethane which becomes soft at body temperature ensuring patient comfort
  • Precisely tapered and siliconised catheter for smooth advancement of the catheter
  • Catheter with X-ray contrast strips for higher patient safety when placement goes wrong
  • Made out of kink resistant material to prevent kinking during placement for a higher patient comfort
  • Less chance of mechanical phlebitis because it is made out of high quality polyurethane so that it becomes softer at body temperature
TRUSTED_SUSTAINABILITY
Especificaciones
Material Polyurethane, Stainless Steel (Needle)
Color Pink
Calibre de aguja 20 G
Sin látex Yes
Longitud (métrico) 32 mm
Tipo de producto IV Catheter
Desechables Single-use
Para utilizar con (equipo) IV Lines
Tipo de conexión Leur Slip
Graduated No
Esterilidad Sterile