Learn More
Saint-Gobain Performance Plastics PureFlo™ Z Series PES Mid Size Capsules
Código de producto. 30428858
Change view
Click to view available options
Tamaño de poro:
0.2 μm
0.45 μm
0.45/0.2 μm
Cantidad:
24 Capsules
30 Capsules
36 Capsules
Tamaño de la unidad:
paquete de 24
paquete de 30
paquete de 36
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tamaño de poro Cantidad unitSize
30428858 0.2 μm 24 Capsules paquete de 24
30428828 0.45 μm 24 Capsules paquete de 24
30428873 0.45/0.2 μm 36 Capsules paquete de 36
30428821 0.2 μm 36 Capsules paquete de 36
30428809 0.2 μm 30 Capsules paquete de 30
30428886 0.45/0.2 μm 24 Capsules paquete de 24
30428803 0.45/0.2 μm 30 Capsules paquete de 30
30428879 0.45 μm 36 Capsules paquete de 36
30428822 0.45 μm 30 Capsules paquete de 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Código de producto. 30428858 Proveedor Saint-Gobain Performance Plastics N.º de proveedor SZVS020ETCTC
Hydrophilic, gamma-irradiatable filtration units designed for single-use sterile assemblies. With superior flow, throughput, broad chemical compatibility and scalable. They are reliable pre- and post-use integrity testing for critical biopharmaceutical processes.
  • PureFlo™ Z Series PES Capsules are designed for single-use sterile assemblies, providing hydrophilic filtration for critical biopharmaceutical processes.
  • Gamma-irradiatable design supports effective sterilization, ensuring compatibility with stringent pharmaceutical requirements.
  • Superior throughput capacity and flow performance enhance operational efficiency in liquid applications.
  • Hydrophilic, asymmetric polyethersulfone (PES) membrane delivers reliable filtration and chemical compatibility across a broad range of substances.
  • Available with a final sterilizing grade 0.1 or 0.2 μm membrane, ensuring effective filtration.
  • Optional built-in ZenFlo™ prefilter enhances performance and efficiency.
  • Carefully selected materials and design enable quick and reliable pre- and post-use integrity testing for critical applications.
  • Scalable size options simplify scale-up throughout all stages of drug development and manufacturing processes.
  • Autoclavable construction ensures flexibility in sterilization methods for optimized usability.
  • Lightweight and compact design integrates seamlessly into biopharmaceutical assemblies without adding bulk.
  • Promotes optimal process efficiency, minimizing downtime and supporting productivity in demanding environments.
TRUSTED_SUSTAINABILITY
Especificaciones
Desechables Single-use
Tipo de filtro Filter Capsule
Material (membrana) Hydrophilic PES
Tamaño de poro 0.2 μm
Esterilidad Non-sterile
Cantidad 24 Capsules
Tipo Mid Size Capsules