Learn More
Kinesis X100 Filtration column TELOS (2 x20µm PE frits)polypropylene, polyethylene 15mL (pack of 100)
Marca:  Kinesis 06479-23
X100 Filtration column TELOS (2 x20µm PE frits)polypropylene, polyethylene 15mL (pack of 100)
Código de producto. 12926089
  • ONLINE EXCLUSIVE PRICE
    199.00€
    166.43€ / paquete de 100
    Ahorro 32.57€
    16% Descuento