Learn More
Kartell SpA Volumetric Flask with Cap
Descripción
Crystal clear volumetric flask creates no meniscus so final readings are easy. Calibrated gravimetrically at 20°C. Thick walls eliminate distortion. Thin necks make final measurement precise. Autoclavable to 170°C. Chemically non-absorbant. No wetting. Stopper included
Especificaciones
Especificaciones
| Capacidad (métrico) | 500 mL |
| Material | PMP |
| Tipo de producto | Volumetric Flask |
| Esterilizables en autoclave | Autoclavable |
| Desechables | Reusable |
| Graduated | No |
| Mouth Style | Threaded |
| Esterilidad | Non-sterile |
| Forma del fondo | Round |
| Tratamiento de superficie | Untreated |