missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT9A Monoclonal Antibody (326711), Alexa Fluor™ 488
Shop All Thermo Scientific Products

Description
In direct ELISAs, no cross-reactivity with recombinant human Wnt-2, -3a, -6, -7a, -7b, recombinant mouse Wnt-1, -2b, -4, -5a, -5b, -8a, -8b, -9b, -10a, -10b, -11, or -16 is observed.
WNT9A is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. WNT9A is likely to signal over only few cell diameters.
Specifications
Specifications
| Antígeno | WNT9A |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 326711 |
| Conjugado | Alexa Fluor 488 |
| Formulación | Buffered saline solution with BSA and 0.1% sodium azide |
| génica | WNT9A |
| N.º de referencia del gen | O14904 |
| Alias de gen | MGC138165; MGC141991; Protein Wnt-14; Protein Wnt-9a; wingless related MMTV integration site 9A; wingless-type MMTV integration site 14; wingless-type MMTV integration site 9A; wingless-type MMTV integration site family member 9A; wingless-type MMTV integration site family, member 14; wingless-type MMTV integration site family, member 9A; Wnt family member 9A; WNT14; wnt-14; Wnt9a |
| Símbolos de los genes | WNT9A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?