Learn More
PeproTech WNT3A Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Descripción
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Wnt-3a. Anti-Human Wnt-3a-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hWNT-3a by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human WNT-3a (500-P251) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hWNT-3a. Western Blot: To detect hWNT-3a by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hWNT-3a is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P251BT-1MG will be provided as 2 x 500 μg
Especificaciones
Especificaciones
| Antígeno | WNT3A |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1-1.0 mg/mL |
| Conjugado | Biotin |
| Formulación | PBS with no preservative |
| génica | WNT3A |
| N.º de referencia del gen | P56704 |
| Alias de gen | MGC119418; MGC119419; MGC119420; Protein Wnt-3a; vestigial tail; vt; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3A; Wnt 3a; Wnt family member 3A; wnt[a]; wnt3; wnt3 l; wnt3 like; Wnt3a; Wnt-3a; WNT3A protein; wnt3l; Zwnt[a] |
| Símbolos de los genes | WNT3A |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.