missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT2B Recombinant Rabbit Monoclonal Antibody (9M6O7)
Descripción
Immunogen Sequence: KAFVDAKEKR LKDARALMNL HNNRCGRTAV RRFLKLECKC HGVSGSCTLR TCWRALSDFR RTGDYLRRRY DGAVQVMATQ DGANFTAARQ GYRRATRTDL V.
WNT2B is a member of the wingless-type MMTV integration site (WNT) family of highly conserved, secreted signaling factors. WNT family members function in a variety of developmental processes including regulation of cell growth and differentiation and are characterized by a WNT-core domain. It may play a role in human development as well as human carcinogenesis. Two alternatively spliced transcript variants exist.
Especificaciones
Especificaciones
| Antígeno | WNT2B |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Recombinant Monoclonal |
| Clon | 9M6O7 |
| Concentración | 0.6 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | WNT2B |
| N.º de referencia del gen | O70283, Q93097 |
| Alias de gen | Protein Wnt-13; Protein Wnt-2b; wingless related MMTV integration site 2b; wingless-type MMTV integration site 2B; wingless-type MMTV integration site family member 2B; wingless-type MMTV integration site family, member 13; wingless-type MMTV integration site family, member 2B; Wnt family member 2B; Wnt13; wnt-13; Wnt2b; XWNT2, Xenopus, homolog of |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?