Learn More
TERUMO Winged Infusion Safety Sets SURFLO™
Descripción
- Made out of kink resistant material to prevent kinking during placement for a higher patient comfort
- The wings are large and flexible for a good grip and secure fixation
- Reduced pressure on the blood vessel wall due to slightly angled wings
- Easy activation of the safety mechanism, fingers tips stay away from the cannula at all times
- Audible sound during activation of the safety mechanism
Especificaciones
Especificaciones
| Tipo de producto | Infusion Set |
| Color | Pink |
| Material | PVC Tubing, Siliconized Needle |
| Calibre de aguja | 18 G |
| Esterilidad | Sterile |
| Sin látex | Yes |
| Longitud del tubo (métrico) | 30 cm |