missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WHSC2 Polyclonal Antibody

Descripción
WHSC2 Polyclonal Antibody for Western Blot, IHC (P)
This gene is expressed ubiquitously with higher levels in fetal than in adult tissues. It encodes a protein sharing 93% sequence identity with the mouse protein. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene is mapped to the 165 kb WHS critical region, and may play a role in the phenotype of the WHS or Pitt-Rogers-Danks syndrome. The encoded protein is found to be capable of reacting with HLA-A2-restricted and tumor-specific cytotoxic T lymphocytes, suggesting a target for use in specific immunotherapy for a large number of cancer patients. This protein has also been shown to be a member of the NELF (negative elongation factor) protein complex that participates in the regulation of RNA polymerase II transcription elongation.
Especificaciones
Especificaciones
| Antígeno | WHSC2 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| génica | NELFA |
| N.º de referencia del gen | Q8BG30, Q9H3P2 |
| Alias de gen | FLJ10442; FLJ25112; LOW QUALITY PROTEIN: negative elongation factor A; mWHSC2; Negative elongation factor A; negative elongation factor complex member A; negative elongation factor complex member A, Whsc2; Nelfa; NELF-A; P/OKcl.15; Whsc2; Whsc2h; Wolf-Hirschhorn syndrome candidate 2; Wolf-Hirschhorn syndrome candidate 2 homolog; Wolf-Hirschhorn syndrome candidate 2 protein |
| Símbolos de los genes | NELFA |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?