Learn More
SHOWA™ White General Purpose Gloves, Model B0610
Descripción
- Designed for optimum freedom of movement, the B0610 has a short wrist cuff.
- The uncoated, seamless nylon liner reduces irritation and allergy risks while maintaining dexterity and tactility.
Especificaciones
Especificaciones
| Coating Material | Uncoated |
| Color | Blanco |
| Desechables | Reusable |
| Hand | Ambidiestro |
| Laboratory/Cleanroom Use | Sí |
| Latex Free | Sí |
| Material | Nylon |
| Tipo de producto | Glove |
| Tamaño | 6 |
| Esterilidad | No estéril |
| Mostrar más |