Learn More
DWK Life Sciences WHEATON™ CompletePAK Sterile ModPAK Sterile Vial Kit
Código de producto. p-7221820 Tienda Wheaton Science Products Productos
Change view
Click to view available options
Volumen (métrico):
2 mL
5 mL
10 mL
Tipo de tapón:
Tapón Lyo
Tapón OmniFlex
Tapón sin revestimiento
Color del tapón:
Azul
Rojo
Tamaño de la unidad:
1 set
Código de producto. Volumen (métrico) Tipo de tapón Color del tapón unitSize
17962581 2 mL Tapón Lyo Azul 1 set
17982651 2 mL Tapón Lyo Rojo 1 set
17952651 2 mL Tapón OmniFlex Azul 1 set
17912661 2 mL Tapón OmniFlex Rojo 1 set
17962651 2 mL Tapón sin revestimiento Azul 1 set
17972651 2 mL Tapón sin revestimiento Rojo 1 set
17922591 5 mL Tapón Lyo Azul 1 set
17932661 5 mL Tapón Lyo Rojo 1 set
17942661 5 mL Tapón OmniFlex Azul 1 set
17992651 5 mL Tapón OmniFlex Rojo 1 set
17912591 5 mL Tapón sin revestimiento Azul 1 set
17902661 5 mL Tapón sin revestimiento Rojo 1 set
17962661 10 mL Tapón Lyo Azul 1 set
17982661 10 mL Tapón Lyo Rojo 1 set
17952571 10 mL Tapón OmniFlex Azul 1 set
17952661 10 mL Tapón OmniFlex Rojo 1 set
17922661 - Tapón sin revestimiento Azul 1 set
17972571 10 mL Tapón sin revestimiento Rojo 1 set
18 options
Código de producto. 17962581
Marca: DWK Life Sciences W991002CPMP
Solo quedan null
Añadir a la cesta
Añadir a la cesta
WHEATON™ CompletePAK is a range of primary packaging components that are stocked ready-to-use and are customizable for most drug applications
WHEATON™ COMPLETEPAK product line is a range of crimp top vials, rubber stoppers and seals that are ready-to-use and are off-the-shelf, which can be customized for any application.
Each kit comes with specific United States Pharmacopeia (USP) certificates showing that the product meets or exceeds critical USP standards that are enforced by the Federal Drug Administration (FDA).
Key Benefits:
  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components
  • Kitted solutions feature equal amounts of RtU components for reduced scrap rates
  • Complete manufacturing and certificate of analysis traceability
  • CompletePAK Amber for light sensitive content packaging
  • CompletePAK ModPAK Bulk configurations
  • 18 month shelf life from date of processing
  • Container Closure Integrity (CCI) certification available upon request
    Materials
  • Vials are designed for parenteral applications and manufactured from low extractable borosilicate glass
  • All stoppers are manufactured from an ultra pure bromobutyl latex free formulation with extremely high chemical
    purity and low gas permeability
  • Omniflex stoppers offer low levels of residual binding to maximize extraction of active pharmaceutical ingredients
  • Omniflex stopper elasticity delivers a more effective barrier to optimize container closure integrity and content
    protection
  • Igloo lyopholized stoppers are suited for biologics and content with sensitivities to silicon
Certifications
USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and lot certified to SAL <10-6, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
TRUSTED_SUSTAINABILITY
Especificaciones
Color del tapón Azul
Color Clear
Diámetro exterior (imperial) 15 mm
Material Borosilicate Glass
Precortados No
Esterilidad Estéril
Desechables Single-use
Volumen (métrico) 2 mL
Para utilizar con (aplicación) Parenteral Preparations and Ready-to-Use Applications
Graduado No
Incluye Vial, tapón y cierre
Material (sello) Aluminum
N.º por caja 1668/Cs.
Modelo Crimp top
Línea de productos Wheaton CompletePAK
Tipo de tapón Tapón Lyo
Tamaño del tapón 13 mm
Marking Spot No
Esterilizables en autoclave Autoclavable
Altura (métrico) 32 mm