Learn More
GE Healthcare Whatman™ Optitran™ Nitrocellulose Membranes
Descripción
- 100% nitrocellulose supported by an inert polyester nonwoven membrane
- Support does not affect transfer conditions or results
- High sensitivity with very low nonspecific binding
- Stringent washing and blocking conditions are not necessary
- Binding capacity: 75 to 90μg/cm2
- Autoclavable using liquid cool cycle
- Also available in circles
Especificaciones
Especificaciones
| Longitud (métrico) | 30 cm |
| Anchura (métrico) | 3 m |
| Capacidad de unión | 75 to 90 μg/cm2 |
| Cantidad | Each |
| Formato | Rollo |
| Material | Nitrocelulosa |
| Tamaño de poro | 0,45 μm |
| Dimensions (LxW) | 30 cm x 3 m |