Learn More
Cytiva Whatman™ Minifold I Slot-Blot System, complete 48-well (acrylic)
Descripción
The Minifold I System consists of four basic components: sample well plate, filter support plate, vacuum plenum, and metal clamping plate. The sample well plate is available in three configurations for producing spots, dots, or slots.
The Minifold I System is compatible with multichannel pipettes. All three plates are interchangeable and can be purchased as accessory plates or in conjunction with a complete system. The tension on the clamping is adjustable, permitting use of a variety of blotting and filtration media.
Minifold I Spot-Blot System, 48 well
Preferred format for densitometric scanning, since slots can be easily quantitated.
Concentrated signal: 6 mm² sample application area results in high signal intensity.
Easy to survey format: 48 samples on your membrane are easier to view than 96 samples.
The Minifold I System is compatible with multichannel pipettes. All three plates are interchangeable and can be purchased as accessory plates or in conjunction with a complete system. The tension on the clamping is adjustable, permitting use of a variety of blotting and filtration media.
Minifold I Spot-Blot System, 48 well
Preferred format for densitometric scanning, since slots can be easily quantitated.
Concentrated signal: 6 mm² sample application area results in high signal intensity.
Easy to survey format: 48 samples on your membrane are easier to view than 96 samples.
Especificaciones
Especificaciones
| Tipo | Sistema de transferencia de ranura Minifold I |
| Material | Acrílico |
| Volumen de la muestra (métrico) | 1000 μL/well |