missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ West Nile Virus Envelope (Domain III) (lineage 1, strain NY99) Monoclonal Antibody (03)
GREENER_CHOICE

Código de producto. 16333704
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16333704 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16333704 Proveedor Invitrogen™ N.º de proveedor MA529988

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for West Nile Virus (WNV) (lineage 1, strain NY99) E/Envelope (Domain III).

West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins which play a major role for WNV entry into target cells. The viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway as delivery of core gene delivery into the striatum of mouse brain and skeletal muscle resulted in cell death and inflammation.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno West Nile Virus Envelope (Domain III) (lineage 1, strain NY99)
Aplicaciones ELISA
Clasificación Monoclonal
Clon 03
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative; pH 6
Alias de gen West Nile envelope; West Nile Virus; WNV envelope
Especie del huésped Mouse
Inmunógeno Recombinant West Nile Virus (WNV) (lineage 1, strain NY99) E/Envelope (Domain III).
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
Especies diana Virus
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.