missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR4 Monoclonal Antibody (OTI4G1), TrueMAB™, OriGene
Descripción
Required for 7-methylguanosine modification of tRNA. This protein is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD). which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes. including cell cycle progression. signal transduction. apoptosis. and gene regulation. This protein is excluded as a candidate for a form of nonsyndromic deafness (DFNB10). but is still a candidate for other disorders mapped to 21q22.3 as well as for the development of Down syndrome phenotypes. Alternative splicing of this gene generates at least 3 transcript variants. two of which diverge at the 3′ UTR and thus encode the same protein products.
Especificaciones
Especificaciones
| Antígeno | WDR4 |
| Aplicaciones | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI4G1 |
| Concentración | 1.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | WDR4 |
| N.º de referencia del gen | P57081 |
| Alias de gen | TRM82, TRMT82 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?