missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wash Buffer 1 (concentrated), Thermo Scientific™
Tienda Thermo Scientific Productos
Click to view available options
Cantidad:
25 mL
Tamaño de la unidad:
25 ml
Description
Thermo Scientific GeneJET Wash Buffer 1 (concentrated) is a component of several GeneJET nucleic acid purification kits and may be purchased separately. Provided in 25 mL aliquots. Please consult kit protocols to confirm catalog numbers for re-ordering the appropriate kit-specific components.
Specifications
Specifications
| Nombre del producto químico o material | Wash buffer |
| Color | Incolora |
| Descripción | Wash Buffer 1, 25mL, Concentrated, Colorless, Solution, 6.8 pH Range, Wash Buffer, Used For Viral RNA/DNA Purification |
| Para utilizar con (aplicación) | Viral RNA/DNA purification; DNA and RNA purification and analysis |
| pH | 6.8 |
| Forma física | Solution |
| Cantidad | 25 mL |
| Formulación | Solution |
| Olor | Odorless |
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction