Learn More
WASF3 Mouse anti-Human, Unconjugated, Clone: 3B7D10, Proteintech
Código de producto. 16880346 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16880346 20 μL 20 microlitros
16870346 150 μL 150 microlitros
2 options
Código de producto. 16880346
Marca: Proteintech 676201IG20UL
Mouse Monoclonal Antibody
The WASF3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WASF3. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The WASF3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
Antígeno WASF3
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence
Clasificación Monoclonal
Clon 3B7D10
Concentración 500 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 10810
Especies diana Human, Mouse, Rat, Pig, Rabbit
Isotype IgG1