missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: HEFPPLFAFP SLSQTDLEEL TYFFPEDHAE DVQQEFFPSW PTPSGLTEYS TLTLCQETLA NSSIGRLCLA FLGKRLDSVI EMCVKDVLLK DDLSWAEAGV AL Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.
Especificaciones
Especificaciones
| Antígeno | VWDE |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | VWDE |
| N.º de referencia del gen | Q8N2E2 |
| Alias de gen | von Willebrand factor D and EGF domain-containing protein; von Willebrand factor D and EGF domains; VWDE |
| Símbolos de los genes | VWDE |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
