missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS45 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12006-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting genes involved in vesicle transport to vacuoles. This gene is a member of the Sec1 domain family, and shows a high degree of sequence similarity to mouse, rat and yeast Vps45. The exact function of this gene is not known, but its high expression in peripheral blood mononuclear cells suggests a role in trafficking proteins, including inflammatory mediators.Especificaciones
| VPS45 | |
| Polyclonal | |
| Unconjugated | |
| VPS45 | |
| h VPS45, H1, H1VPS45, hlVps45, VPS45, VPS45A, VPS45B, VPS54A, VSP45, VSP45A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11311, 22365, 64516 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08700, P97390, Q9NRW7 | |
| Vps45 | |
| VPS45 Fusion Protein Ag2658 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido