missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS25 Polyclonal Antibody

Goat Polyclonal Antibody
Marca: Invitrogen™ PA518464
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
VPS25 (Vacuolar Protein Sorting 25 Homolog) is a Protein Coding gene. Among its related pathways are Delta508-CFTR traffic / ER-to-Golgi in CF and Budding and maturation of HIV virion. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and protein N-terminus binding.
Especificaciones
| VPS25 | |
| Polyclonal | |
| Unconjugated | |
| VPS25 | |
| 1110020N13Rik; AW107467; D11Wsu68e; dermal papilla-derived protein 9; DERP9; Eap20; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; FAP20; HGNC:28122; hVps25; MGC10540; vacuolar protein sorting 25 (yeast); vacuolar protein sorting 25 homolog; vacuolar protein sorting 25 homolog (S. cerevisiae); vacuolar protein-sorting-associated protein 25; VPS25 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 681059, 84313 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P0C0A1, Q9BRG1 | |
| VPS25 | |
| Synthetic peptide sequence (QPNVDTRQKQ) corresponding to the internal amino acids of VPS25 (aa 20-29). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction