missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vitronectin Monoclonal Antibody (HV2)
GREENER_CHOICE

Código de producto. 11514162
Change view
Click to view available options
Cantidad:
400 μL
Tamaño de la unidad:
200 microgramos
Código de producto. Cantidad unitSize
11514162 400 μL 200 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11514162

Marca: Invitrogen™ CSI0030202

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

CSI 003-02-02 is highly specific for vitronectin. There is no evidence for cross-reactivity with other connective tissue proteins (fibronectin, elastin, collagen, laminin). CSI 003-02-02 is human-specific (no reactivity with cat, dog, cow, sheep, goat, pig, rabbit, horse). CSI 003-02-02 has been successfully used in ELISA, immunohistochemistry, and Western blotting techniques. CSI 003-02-02 can be used to purify vitronectin from human plasma by affinity chromatography. It can also be used to quantitatively affinity-deplete human plasma or serum of vitronectin. It partially denatures vitronectin upon antibody binding. It binds equally well to native and denatured vitronectin and can be used to quantitate vitronectin in human plasma in a sandwich ELISA with CSI 003-02-02. It does not interfere with the binding of any known vitronectin ligands. The epitope of CSI 003-02-02 is located in the connecting region. Binding can be competed selectively by a peptide comprising aa 121-133. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

VTN is a member of the pexin family. This protein is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. The protein is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Vitronectin
Aplicaciones ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon HV2
Concentración 1.04 mg/mL
Conjugado Unconjugated
Formulación PBS with 15mM sodium azide; pH 7.4
génica VTN
N.º de referencia del gen P04004
Alias de gen Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN
Símbolos de los genes VTN
Especie del huésped Mouse
Inmunógeno Human vitronectin purified from plasma by heparin-affinity chromatography.
Método de purificación Protein A
Cantidad 400 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7448
Especies diana Human
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.