Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™
Código de producto. 18712007
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
18712007 100 μg 100 microgramos
18734217 25 μg 25 microgramos
2 options
Código de producto. 18712007
Marca: R&D Systems MAB11188100
Mouse Monoclonal Antibody
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno HIV-1 Gag p24
Aplicaciones Western Blot, ELISA
Clasificación Monoclonal
Clon 1042960
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, ELISA
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen Q77YG1_9HIV1
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant viral HIV-1 Gag p24, Pro149-Gly354, Accession # Q77YG1_9HIV1
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 155030
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Virus
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1