Learn More
Invitrogen™ Vimentin Monoclonal Antibody (VI-10)
GREENER_CHOICE
Mouse Monoclonal Antibody
Marca:  Invitrogen™ MA110459
Código de producto. 12372453
  • 445.00€ / 100 microgramos
Descripción
This antibody reacts with vimentin, a 57 kDa intermediate filament intracellular protein expressed in variety of mesenchymal and mesodermal cell types. Immunocytochemistry: Staining technique: (a) fix cells for 10 min in methanol at -20°C and for 6 min in acetone at -20°C; (b) fix cells directly in methanol for 10 min at -20°C or in acetone for 10 min at -20°C.
Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Vimentin
Monoclonal
1 mg/mL
TBS with 15mM sodium azide; pH 8
P02543, P08670, P09654, P20152, P31000
Vim
not available.
100 μg
Primary
Human, Mouse, Rat, Pig, Chicken
Antibody
IgM
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
VI-10
Unconjugated
Vim
cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus
Mouse
sequential chromatography
RUO
100522394, 22352, 420519, 7431, 81818
4°C, do not freeze
Liquid
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones