missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Monoclonal Antibody (SP20), Invitrogen™
Descripción
MA5-14564 targets Vimentin in IHC (P) , WB, IF and ICC applications and shows reactivity with Human, Primate, Mouse and Rat samples. The MA5-14564 immunogen is recombit protein encoding human vimentin.
This gene encodes a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein encoded by this gene is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a domit, pulverulent cataract.
Especificaciones
Especificaciones
| Antígeno | Vimentin |
| Aplicaciones | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | SP20 |
| Conjugado | Unconjugated |
| Formulación | tissue culture supernatant diluted in TBS with 1% BSA and 0.1% sodium azide |
| génica | VIM |
| N.º de referencia del gen | P08670 |
| Alias de gen | VIM, FLJ36605 |
| Símbolos de los genes | VIM |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?