Descripción
- Durable microfiber bag securely contains beads eliminating the risk of leakage and is unaffected by moisture
- Indicator dot changes from blue when dry to pink when moist to easily identify when recharging is needed
- Saturated gel can be regenerated by microwaving bag for 6 minutes
- May be regenerated repeatedly without degradation
- Arrives individually sealed in moisture-proof pouch
- Available in multiple sizes to fit most desiccators
Especificaciones
Especificaciones
| Diámetro exterior (imperial) | 6 cm |
| Tipo de producto | Desiccant Bag |
| Descripción | Durable Microfiber Pouch |
| Material | Silica Gel |
| Altura (métrico) | 25 cm |
| Altura (imperial) | 9.75 in. |
| Anchura (métrico) | 6 cm |
| Anchura (imperial) | 2.25 in. |
| Longitud (métrico) | 15 cm |
| Longitud (imperial) | 6 in. |
| Mostrar más |