Learn More
Human IgG2 Antibody, R&D Systems™
Descripción
Human IgG2 Monoclonal specifically detects Human IgG2 in Human samples. It is validated for Western Blot, ELISA.
Especificaciones
Especificaciones
| Antígeno | Human IgG2 |
| Aplicaciones | Western Blot, ELISA |
| Clasificación | Monoclonal |
| Clon | 2348B |
| Conjugado | Unconjugated |
| Dilución | Western Blot 2 μg;/mL, ELISA |
| Formulación | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Alias de gen | Ig gamma-2 chain C region, IGHG2, immunoglobulin heavy constant gamma 2 (G2m marker) |
| Especie del huésped | Rabbit |
| Inmunógeno | Human IgG2 protein |
| Mostrar más |