Learn More
Human PKM1/2 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18760517
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
Código de producto. Cantidad unitSize
18760517 25 μg 25 microgramos
18702097 100 μg 100 microgramos
2 options
Código de producto. 18760517
Marca: R&D Systems MAB72443SP
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Mouse Monoclonal Antibody
PKM2 Monoclonal specifically detects PKM2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PKM2
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon 945103
Conjugado Unconjugated
Dilución Western Blot 0.25-0.5 ug/mL, Immunocytochemistry 5-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen P14618
Alias de gen CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP3, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, p58, PK, muscle type, PK2, PK3, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK
Especie del huésped Mouse
Inmunógeno E.coli-derived recombinant human PKM2, Ser434-Pro531, Accession No. P14618
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Primario o secundario Primary
ID de gen (Entrez) 5315
Especificidad de la prueba Detects human PKM2 in direct ELISAs.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Formulario Supernatant
Isotype IgG2a