Learn More
Human IL-17/IL-17A Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18755239
Change view
Click to view available options
Cantidad:
1 mg
100 μg
25 μg
500 ug
Tamaño de la unidad:
1 miligramo
100 microgramos
25 microgramos
500 microgramos
Código de producto. Cantidad unitSize
18755239 100 μg 100 microgramos
18754639 25 μg 25 microgramos
18188810 1 mg 1 miligramo
18138820 500 ug 500 microgramos
4 options
Código de producto. 18755239
Marca: R&D Systems MAB3171R100
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Mouse Monoclonal Antibody
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IL-17/IL-17A
Aplicaciones Western Blot, Flow Cytometry
Clasificación Monoclonal
Clon 41802R
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Formulación Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen Q16552
Alias de gen CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Especie del huésped Mouse
Inmunógeno E.coli-derived recombinant human IL-17 Ile20-Ala155 Accession # Q16552
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3605
Especificidad de la prueba Detects human IL-17 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) IL-17A/IL-17F heterodimer and no cross-reactivity with rhIL-17F, recombinant mouse IL-17, and recombinant rat IL-17A is observed.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1