Learn More
Human Notch-1 Intracellular Domain Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18727177
Change view
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
25 microgramos
Código de producto. Cantidad unitSize
18727177 25 μg 25 microgramos
1 options
Código de producto. 18727177
Marca: R&D Systems AF3647SP
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Sheep Polyclonal Antibody has been used in 3 publications
Notch-1 Polyclonal specifically detects Notch-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Notch-1
Aplicaciones ChIP Assay, CyTOF, Flow Cytometry, Immunocytochemistry
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P46531
Alias de gen EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1
Símbolos de los genes NOTCH1
Especie del huésped Sheep
Inmunógeno E. coli-derived recombinant human Notch-1 Gly2428-Lys2556 Accession # P46531
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4851
Especificidad de la prueba Detects the intracellular domain (ICD) of human Notch-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Notch-2 ICD, rhNotch-3 ICD, and rhNotch-4 ICD is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG