Learn More
Human/Rat PKM1/2 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18701959
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18701959 100 μg 100 microgramos
18728468 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18701959 Proveedor R&D Systems N.º de proveedor MAB7244
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Mouse Monoclonal Antibody
PKM2 Monoclonal specifically detects PKM2 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Knockout Validated.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PKM2
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown
Clasificación Monoclonal
Clon 937801
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P14618
Alias de gen CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP3, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, p58, PK, muscle type, PK2, PK3, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK
Símbolos de los genes PKM
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human PKM2 Ser434-Pro531 Accession # P14618
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5315
Especificidad de la prueba Detects human PKM2 in direct ELISAs and Western blots.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human, Rat
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2b